missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide, 99% (metals basis)

Product Code. 11369498
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369498 2 kg 2kg
11359498 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369498 Supplier Thermo Scientific Alfa Aesar Supplier No. 043332.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 409-21-2 | CSi | 40.10 g/mol

Silicon carbide powder is used as an abrasive for such as grinding wheels, whetstone, grinding wheel and sand tiles. Silicon carbide is used to produce epitaxial grapheme by graphitization at high temperatures. It is also acts asthe metallurgical deoxidizer material. Four main application areas of SiC: functional ceramics, advanced refractory, abrasives and metallurgical materials. High purity single crystal, is used to manufacture semiconductors, manufacture of silicon carbide fibers. Silicon carbide is very hard, with excellent thermal conductivity, as a semiconductor and high temperature resistant to oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilanylium
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black
Physical Form Powder
Quantity 2 kg
Linear Formula SiC
Merck Index 14,8492
Solubility Information It is soluble in fused potassium hydroxide,and molten iron.It is insoluble in water (0.01mg/ml) & ethanol and acid.
Formula Weight 40.1
Percent Purity 99%
Odor Odorless
Density 3.16 g/mL
Assay (metals basis)
Chemical Name or Material Silicon carbide
Show More Show Less

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.