missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide, 600 grinding compound, 2oz (57g), Thermo Scientific™

Product Code. 15441567
Change view
Click to view available options
Quantity:
57 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441567 57 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15441567 Supplier Thermo Scientific Alfa Aesar Supplier No. 039801.LA
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grinding compound for polishing IR crystal windows

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 409-21-2
Quantity 57 g
Molecular Formula CSi
MDL Number MFCD00049531
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES [C-]#[Si+]
IUPAC Name methanidylidynesilanylium
Molecular Weight (g/mol) 40.10
PubChem CID 9863
ChEBI CHEBI:29390
Formula Weight 40.1
Odor Odorless
Chemical Name or Material Silicon carbide, 600 grinding compound
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.