missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicic Acid, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
674.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.10kg |
Quantity | 500 g |
Physical Form | Powder |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Silicic Acid, n-Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'