missing translation for 'onlineSavingsMsg'
Learn More

Silicic acid hydrate

Product Code. p-4408646
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11387578

Brand: Thermo Scientific Alfa Aesar 033270.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1343-98-2 | O2Si | 60.08 g/mol

Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-98-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym orthosilicic acid, monosilicic acid, tetrahydroxysilane, h4sio4, silicon tetrahydroxide, silicasol, kieselsaeure, si oh 4, unii-623b93yabh, orthosilicicacid
PubChem CID 14942
ChEBI CHEBI:26675
IUPAC Name silicic acid
SMILES O=[Si]=O

Specifications

Appearance White powder
Color White
Quantity 100 g
Physical Form -100 Mesh Powder
Formula Weight 60.09 (Anhydrous)
Chemical Name or Material Silicic acid hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.