missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicagel, functionalized, (cyclohexylcarbodiimido)propyl, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| Quantity | 5 g |
| Formula Weight | 60.08 |
| Chemical Name or Material | Silicagel, Functionalized, (cyclohexylcarbodiimido)propyl, ca. 0.9mmol/g, particle size: 40-63 micron |
Safety and Handling
Product Identifier
- Silicagel, functionalized, (cyclohexylcarbodiimido)propyl
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?