missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ Silica Ultra Low Range Bulk Reagent Set
Specifications
Specifications
| Analytes | Silica (SiO2) |
| Format | Bottle (Amino Acid F reagent) and Dropper Bottles |
| For Use With (Application) | Ultra low range silica determination |
| For Use With (Equipment) | Colorimeters (All Hach models except DR 900, DR/800 Series, PC II) |
| No. of Tests | 100 |
| Quantity | 1 Set |
| Range | 3 to 1000μg/L SiO2 |
| Test Method | Heteropoly blue |
| Type | Reagent |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?