missing translation for 'onlineSavingsMsg'
Learn More

Draeger™ Type B Silica Gel Tubes

Product Code. 10170252
Change view
Click to view available options
For Use With (Equipment):
Accuro Drager Gas Detector Pump
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10170252 Accuro Drager Gas Detector Pump Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10170252 Supplier Draeger™ Supplier No. 6733021
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sorbent tubes using activated charcoal or silica gel to capture airborne contaminants for later laboratory analysis.

  • Typically sufficient for sampling in the range of limit value measurements
  • After sampling, the tube must be sealed with the polyethylene supplied caps
  • Length of 125mm, diameter of 7mm
  • Supplied in packs of 10

Specifications

Certifications/Compliance BGIA, DFG, NIOSH, OSHA and HSE Certified
For Use With (Equipment) Accuro Drager Gas Detector Pump
Quantity 10/Pk.
Sample Type Silica gel
Tube Type Silica Gel Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.