missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica gel, TLC high purity grade, without binder, Thermo Scientific Chemicals
For TLC Plate Preparation
£17.10 - £631.00
Chemical Identifiers
CAS | 63231-67-4 |
---|---|
Molecular Formula | H2O3Si |
Molecular Weight (g/mol) | 78.098 |
MDL Number | MFCD00011232 |
InChI Key | IJKVHSBPTUYDLN-UHFFFAOYSA-N |
Synonym | metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid |
PubChem CID | 14768 |
IUPAC Name | dihydroxy(oxo)silane |
SMILES | O[Si](=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11335557
|
Thermo Scientific Alfa Aesar
042756.18 |
50 g | |||||||
11315557
|
Thermo Scientific Alfa Aesar
042756.30 |
250 g |
£59.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11345557
|
Thermo Scientific Alfa Aesar
042756.A1 |
1 kg | |||||||
11325557
|
Thermo Scientific Alfa Aesar
042756.A7 |
5 kg |
£631.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silica gel, TLC high purity grade, without binder is employed for thin layer chromatography plate preparation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63231-67-4 | |
78.098 | |
IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
14768 | |
O[Si](=O)O |
H2O3Si | |
MFCD00011232 | |
metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid | |
dihydroxy(oxo)silane |
Specifications
63231-67-4 | |
Powder | |
MFCD00011232 | |
Soluble in alkalies. Insoluble in water. | |
O[Si](=O)O | |
78.098 | |
TLC High Purity | |
Silica gel |
White | |
H2O3Si | |
metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid | |
IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
dihydroxy(oxo)silane | |
14768 | |
Odorless |
Safety and Handling
H373: May cause damage to organs through prolonged or repeated exposure.
missing translation for 'einecsNumber' : 231-545-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only