missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Rubin, Honeywell Fluka™

Product Code. p-7120754 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 15657350

Brand: Honeywell-Fluka 858151KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Drying agent, with moisture indicator, cobalt free

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112926-00-8
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Color Red
Melting Point 1600°C
Physical Form Granular
pH 9.0
Material Silica Gel
Quantity 1 kg
UN Number NONH for all modes of transport
Odor Odorless
Packaging Poly Bottle
Chemical Name or Material Silica gel Rubin
Show More Show Less
Product Identifier
  • Silica gel Rubin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.