missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Rubin, Solstice

Product Code. 15667350
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15667350 2.5 kg Poly Bottle 2.5kg
15657350 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15667350 Supplier Honeywell-Fluka Supplier No. 858152.5KG
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Drying agent, with moisture indicator, cobalt free

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112926-00-8
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Color Red
Melting Point 1600°C
Physical Form Granular
pH 9.0
Material Silica Gel
Quantity 2.5 kg
UN Number NONH for all modes of transport
Odor Odorless
Packaging Poly Bottle
Chemical Name or Material Silica gel Rubin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.