missing translation for 'onlineSavingsMsg'
Learn More

Silica gel orange, Honeywell™

Product Code. 16645733 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.50kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16645733 500 g 500g
16625733 1 kg 1kg
16635733 2.5 kg 2.50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16645733 Supplier Honeywell Research Chemicals Supplier No. 10087H500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Granulate (0.2-1 mm)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Beads
CAS 7631-86-9, 7783-83-7
CAS Min % ≥5.00%, ≥90.00%
CAS Max % <10.00%, ≤100.00%
Quantity 500 g
Purity Grade Notes Silica gel
Loss on Drying Max. 2 %
Packaging Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.