missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, large pore, 99.5%

Product Code. 11433770
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433770 2 kg 2kg
11423770 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433770 Supplier Thermo Scientific Alfa Aesar Supplier No. 089385.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol

A useful product for column chromatography (HPLC). Used as cat litter, food additive, water filtration and humidity indicator. Also a reliable substance to preserve field-collected leafs for molecular studies for variation in DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Color White
Physical Form 56 Micron,S.A. 500m
Material Silica Gel
Quantity 2 kg
Particle Size 56 Micron
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Percent Purity 99.5%
Surface Area 500m2/g
Chemical Name or Material Silica gel, Large pore, P.V. 0.75cc/g

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.