missing translation for 'onlineSavingsMsg'
Learn More

Silica gel desiccant, indicating, ACS

Product Code. 11430271
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430271 2.5 kg 2.5kg
11343958 100 g 100g
11333958 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430271 Supplier Thermo Scientific Alfa Aesar Supplier No. 040381.A4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Color Blue
Physical Form Granular
Material Silica Gel
Quantity 2.5 kg
Odor Odorless
Solubility Information Soluble in alkalies. Insoluble in water.
Sensitivity Hygroscopic
Grade ACS
Chemical Name or Material Silica gel desiccant, For analysis ACS
Can I regenerate Silica gel desiccant, indicating, ACS?

Yes, this silica gel desiccant can be regenerated. This is achieved by heating in an oven at ~130 degrees C for 3 hours, or until the product returns to a blue color.

What color change indicates the Silica gel desiccant, indicating, ACS has reached moisture capacity?

The silica gel desiccant blue indicator spheres turn pink when they reach moisture capacity.

compliance-icons
Product Identifier
  • Silica gel desiccant, indicating, ACS
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Cobaltous chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.