missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for chromatography, 0.200-0.500 mm, 60 A

Product Code. p-70228 Shop All Thermo Scientific Chemicals Products
Click to view available options
Packaging:
Plastic Bottle
Plastic Drum
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 10256770

Brand: Thermo Scientific Chemicals 240380010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 7631-86-9 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications:

For column chromatography

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9

Specifications

Pore Size 60 Å
Packaging Plastic Bottle
Quantity 1 kg
Type Silica Gel
Material Silica Gel
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.