missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for chromatography, 0.075-0.250 mm, 150 A

Product Code. 10316451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316451 1 kg Glass bottle 1kg
10460983 250 g Glass bottle 250g
10646994 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10316451 Supplier Thermo Scientific Acros Supplier No. 419280010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7631-86-9 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color White
Physical Form Very Fine Granular Powder
Packaging Glass bottle
Particle Size Nominal particle size: >60 mesh: <7.5%, <200 mesh: <10.0%
Specific Surface Area 275 to 375 m2/g (BET)
Quantity 1 kg
Merck Index 15, 8632
Solubility Information Solubility in water: insoluble.
IUPAC Name dioxosilane
Formula Weight 60.08
Chemical Name or Material Silica gel, For column chromatography
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.