missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, for chromatography, 0.035-0.070 mm, 90 A, Thermo Scientific Chemicals

Product Code. 10721804
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Silicon dioxide
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 250 g
Packaging Plastic Bottle
Solubility Information Solubility in water: insoluble.
Formula Weight 60.08
Chemical Name or Material Silica gel, For chromatography, 0.035-0.070mm, 90Å

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'