missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, C8-RP, 12%C, ca. 1.2 mmol/g, part. size 40-63 micron

Product Code. 10340484 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10340484 250 g Plastic Bottle 250g
10540352 50 g Plastic Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10340484 Supplier Thermo Scientific Acros Supplier No. 360142500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00148339
Synonym Octylsilicate

Specifications

Color White
Physical Form Powder
Material Silica Gel
Quantity 250 g
Packaging Plastic Bottle
Particle Size 40 to 63 Micron
Chemical Name or Material Silica gel, C8-RP, 12%C, ca. 1.2 mmol/g, part. size 40-63 micron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.