missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, C18-RP, 23%C, approx. 1 mmol/g, part. size: 40-63 micron

Product Code. 10733603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10733603 50 g 50g
10649863 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10733603 Supplier Thermo Scientific Chemicals Supplier No. 360110500
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography

TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
Quantity 50 g
Material Silica Gel
What is the pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 μm (Cat. No. 360110500, 360112500) in Angstroms?

The pore size of Silica gel, C18-RP, 23%C, ca.1.2 mmol/g, part. size 40-63 µm (Cat. No. 360110500, 360112500) is 60 Angstroms.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.