missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, 80 to 200 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15558124 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15558124 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558124 Supplier Avantor J.T.Baker Supplier No. 340401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    Suitable for Column Chromatography and the Scale-up of TLC Procedures

Specifications

CAS 63231-67-4
Material Silica Gel
Quantity 500 g
Molecular Formula H2O3Si
Mesh Size 80 to 200 Mesh
Packaging Wide Mouth Amber Glass Bottle
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
SMILES O[Si](=O)O
IUPAC Name dihydroxy(oxo)silane
Molecular Weight (g/mol) 78.098
PubChem CID 14768
Specific Gravity 1L = 2.10kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Silica Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.