missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.019-0.037mm (400-600 mesh)

Product Code. 11385507
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385507 5 kg 5kg
11305517 50 g 50g
11315517 250 g 250g
11395507 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11385507 Supplier Thermo Scientific Alfa Aesar Supplier No. 042729.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AA4272918 | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Column chromatography with elevated pressureSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Physical Form Powder
Color White
Quantity 5 kg
Odor Odorless
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Chemical Name or Material Silica gel 60

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.