Learn More
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Description
Column chromatographySilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder, -230+400 Mesh,S.A. 500-600m |
| Mesh Size | 230-400 Mesh |
| Odor | Odorless |
| Assay Percent Range | 99.5% (metals basis) |
| Color | White |
| Quantity | 250 g |
| Solubility Information | Soluble in hydrofluoric acid. Insoluble in water. |
| IUPAC Name | dioxosilane |
| Chemical Name or Material | Silica gel 60 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.