missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)

Product Code. p-4408672
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399168 50 g 50g
11329178 250 g 250g
11309178 1 kg 1kg
11319178 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11399168 Supplier Thermo Scientific Alfa Aesar Supplier No. 042570.18
Add to basket
This item is not returnable. View return policy

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Column chromatographySilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Physical Form Powder, -230+400 Mesh,S.A. 500-600m
Mesh Size 230-400 Mesh
Odor Odorless
Assay Percent Range 99.5% (metals basis)
Color White
Quantity 50 g
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
IUPAC Name dioxosilane
Chemical Name or Material Silica gel 60

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.