missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Silica G/UV254 Adsorbents

Brand:  Macherey-Nagel™ 816320.5

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10450712

  • £303.00 / 5kg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

5 kg
2 to 20 μm
0.75 mL/g
Silica G/UV254 Adsorbent
Silica
60 Å
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers