missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Silica Chromabond Diamino Adsorbent

Brand:  Macherey-Nagel™ 730653

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10743324

  • £369.00 / 100g
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

100 g
45 μm
2 to 8
Chromabond Diamino Adsorbent
Silica
60 Å
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers