missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Silica Chromabond C18 Adsorbent

Product Code. 10062861 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10062861 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10062861 Supplier Macherey-Nagel™ Supplier No. 730602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 100 g
For Use With (Application) Non-Polar Compounds, Pesticides
Product Type Chromabond C18 Adsorbent
Particle Size 45 μm
Material Silica
pH Range 2 to 8
Pore Size 60 Å
Surface Area (Metric) 500 m2/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.