missing translation for 'onlineSavingsMsg'
Learn More

Silica 63-100μm, 60Å, EcoChrom™, MP Biomedicals™

Product Code. 11383599 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
Product Code. Quantity unitSize
11383599 25 kg 25kg
1 options
This item is not returnable. View return policy

Product Code. 11383599

Brand: MP Biomedicals 0204660

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Chromatography; Chemical Synthesis

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 kg
Content And Storage Store at Room Temperature.
Type Silica
Physical Form Powder
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Density 2g/cm3 at 20°C
CAS 7631-86-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.