missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica 32-63μm, 60A, EcoChrom™, MP Biomedicals™
For Chromatography; Chemical Synthesis
£172.78 - £2438.00
Specifications
Content And Storage | Store at Room Temperature. |
---|---|
Type | Silica |
Physical Form | Powder |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
Density | 2g/cm3 at 20°C |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
11363569
|
MP Biomedicals
0202827 |
2.5kg |
£172.78
2.50kg |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11333549
|
MP Biomedicals
0202756 |
15kg |
£2438.00
15kg |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11353569
|
MP Biomedicals
0202826 |
25kg |
£1552.90
25kg |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at Room Temperature. | |
Powder | |
2g/cm3 at 20°C |
Silica | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
7631-86-9 |