missing translation for 'onlineSavingsMsg'
Learn More

Silica, 200 to 500 μm, 60A, MP

Product Code. 17837044 Shop All MP Biomedicals Products
Click to view available options
:
MP Silica, 200 to 500 μm, 60A, 500 g
This item is not returnable. View return policy

Product Code. 17837044

Brand: MP Biomedicals 0202811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EcoChrom™ Silica, the chemical compound silicon dioxide, also known as silica (from the Latin silex), is an oxide of silicon with the chemical formula SiO2 with a mean pore size of 60Å and particles size of 200 to 500 μm.

MP silica are free of particles outside the standard range.

The irregular shape of MP Silica:

  • Permits the easy packing of column permeability and plate number
  • Allows easy switching from one sieve cut to another, offering an easy way to change the chromatographic methods
  • Has been known for its hardness since ancient times

Silica is most commonly found in nature:

  • As sand or quartz
  • As in the cell walls of diatoms

Application Notes:

  • MP EcoChrom™ SILICA is used as a suspending and thickening agent in pharmaceuticals
  • A silica-based aerogel was used in the Stardust spacecraft to collect extraterrestrial particles
  • Used in the extraction of DNA and RNA due to its ability to bind to the nucleic acids under the presence of chaotropes
  • As hydrophobic silica it is used as a defoamer component
  • In hydrated form, it is used in toothpaste as a hard abrasive to remove tooth plaque.

SiO2 has a number of distinct crystalline forms (polymorphs) in addition to amorphous forms. In the majority of silicates, the Si atom shows tetrahedral coordination, with 4 oxygen atoms surrounding a central Si atom.

Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Chromatography
  • Chemical Synthesis
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2230°C (Lit.)
Melting Point 1710°C (Lit.)
CAS 7631-86-9
pH 7.0 (Lit.)
Physical Form Powder
Quantity 500 g
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Density 0.5 g/mL
Chemical Name or Material Silicon dioxide
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.