missing translation for 'onlineSavingsMsg'
Learn More

Silibinin, MedChemExpress

Product Code. 16446306
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446306 100 mg 100mg
16491696 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446306 Supplier Medchem Express Supplier No. HY13748

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Silibinin (Silibinin A), an effective anti-cancer and chemopreventive agent, has been shown to exert multiple effects on cancer cells, including inhibition of both cell proliferation and migration.

Silibinin (Silibinin A), an effective anti-cancer and chemopreventive agent, has been shown to exert multiple effects on cancer cells, including inhibition of both cell proliferation and migration.

Specifications

Chemical Name or Material Silibinin
CAS 22888-70-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H22O10
Quantity 100 mg
Synonym Silibinin A Silymarin I
Solubility Information DMSO : 250 mg/mL (518.20 mM; Need ultrasonic)
SMILES O=C1[C@H](O)[C@@H](C2=CC=C(O[C@H](CO)[C@@H](C3=CC=C(O)C(OC)=C3)O4)C4=C2)OC5=CC(O)=CC(O)=C15
Molecular Weight (g/mol) 482.44
Formula Weight 482.44
Percent Purity 99.47%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.