Learn More
Thermo Scientific™ Silencer™ siRNA Library, In Plate
Product Code. 10320065
Change view
Click to view available options
Quantity:
2 nmol
4 nmol
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10320065 4 nmol Each
10764017 2 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 10320065 Supplier Thermo Scientific™ Supplier No. AM81800
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
For Research Use Only. All usage must comply with product instructions.
Silencer™ siRNA Libraries feature combinations of Silencer™ Pre-designed siRNAs, and when available, Validated siRNAs in 96-well or 384-well plates.
  • Silencer™ siRNA Libraries feature combinations of Silencer™ Pre-designed siRNAs
  • When available, Silencer™ Validated siRNAs, in 96-well or 384-well plates.
TRUSTED_SUSTAINABILITY
Specifications
Product Type siRNA Library
Quantity 4 nmol
Format 96- or 384-well Plate
RNAi Type siRNA