missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Select Pre-Designed siRNA

Product Code. 10147724
Change view
Click to view available options
Purity:
HPLC
HPLC purified
In Vivo
Standard
Standard purity
Quantity:
1 μmol
1 nmol
10 μmol
20 nmol
20 nmol (Inventoried)
250 nmol
40 nmol
40 nmol (Inventoried)
5 nmol
5 nmol (Inventoried)
Unit Size:
1nmol
5nmol
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Purity Quantity unitSize
10147724 Standard purity 5 nmol Each
10714037 Standard 1 nmol 1nmol
30245143 Standard purity 5 nmol 5nmol
10782097 Standard 5 nmol (Inventoried) 5nmol
10157724 Standard purity 20 nmol Each
10584615 Standard 20 nmol (Inventoried) Each
10704617 Standard purity 40 nmol Each
10554045 Standard 40 nmol (Inventoried) Each
15581084 HPLC 1 μmol Each
15551084 HPLC 10 μmol Each
10045664 HPLC purified 20 nmol Each
10055664 HPLC 40 nmol Each
10330995 HPLC purified 250 nmol Each
15551064 In Vivo 10 μmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10147724 Supplier Ambion™ Supplier No. 4390771 / W564316928

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry.

Silencer Select Predesigned siRNAs offer researchers a powerful and reliable tool for gene silencing applications. Designed with advanced algorithms to ensure high specificity and efficiency, these siRNAs offer a reliable and powerful solution for gene silencing applications. Chemically modified with locked-nucleic acid (LNA) modifications to enhance stability and reduce off-target effects, Silencer Select Predesigned siRNAs deliver consistent and reproducible results across a wide range of experimental conditions.

Key features include:

  • Precise silencing—minimize off-target effects with the combination of algorithm design and LNA chemical modifications
  • Enhanced potency—achieve effective gene silencing with lower siRNA concentrations
  • Guaranteed silencing—two out of two predesigned siRNAs will result in gene knockdown
  • Comprehensive support—access detailed protocols and expert technical support

Optimized design for enhanced specificity

Our internally developed algorithm has been fine-tuned to create siRNAs with sequences that maximize on-target effects while minimizing off-target interactions. This optimized design approach significantly reduces the risk of unintended gene silencing, thereby increasing the reliability and interpretability of experimental results. The specificity of Silencer Select siRNAs allows researchers to confidently attribute observed phenotypes to the silencing of their gene of interest.

Effective silencing at lower concentrations

A key advantage of Silencer Select siRNAs is their ability to achieve effective silencing at lower siRNA concentrations. This not only reduces experimental costs but also minimizes the potential for off-target effects that can occur at higher doses. Maintaining high knockdown efficiency at reduced concentrations highlights the potency and precision of our siRNA design.

Streamlined workflow for rapid results

Utilizing our gene knockdown technology is straightforward and efficient. Simply order an siRNA for the desired gene target, deliver it to the cells, and detect knockdown efficiency and validate the resulting phenotype. Take advantage of our validated siRNA designs for better confidence in your experiments. This streamlined process allows for rapid experimental progress and accelerated discovery in gene function studies.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains dried siRNA (5 nmol) and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °.
Description Silencer™ Select Pre-Designed siRNA, 5nmol, Standard Purity, For RNAi, epigenetics and Non-Coding RNA research, RNAi, siRNA
RNAi Type siRNA
Final Product Type Silencer™ Select pre-designed siRNA
For Use With (Application) RNAi, epigenetics and non-coding RNA research, RNAi, siRNA
Product Line Silencer Select, Ambion
Product Type Silencer™ Select pre-designed siRNA
Purity Standard purity
Quantity 5 nmol
Sample Size 5 nmol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.