Learn More
Ambion™ Silencer™ Select Custom Designed siRNA
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
Brand: Ambion™ 4399665
148.74 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00450kg
Description
- Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
- Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below -20°C. | |
siRNA | |
Silencer™ Select | |
Standard | |
Ambient |
GeneAssist - Custom Designed Silencer™ Select siRNA | |
Knocking down gene expression in order to study protein function in a wide range of cell types | |
Custom Designed siRNA | |
5 nmol | |
Upon receipt, store in a non-frost-free freezer at or below -20°C |
For Research Use Only. Not for use in diagnostic procedures.