missing translation for 'onlineSavingsMsg'
Learn More

Sildenafil N-Oxide, TRC

Product Code. 17994015
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994015 250 mg 250mg
17921355 50 mg 50mg
17954975 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994015 Supplier Toronto Research Chemicals Supplier No. TRCS435035250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1094598-75-0
Molecular Formula C22 H30 N6 O5 S
Molecular Weight (g/mol) 490.58
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-methyl-4-oxido-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, 5-[2-Ethoxy-5-[(4-methyl-4-oxido-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, Sildenafil-N-oxide, Sildenafil N-Oxide, 5-[2-Ethoxy-5-[(4-methyl-4-oxidopiperazin-1-yl)sulfonyl]phenyl]-1-methyl-3-propyl-6,7-dihydro-1Hpyrazolo[4,3-d]pyrimidin-7-one
IUPAC Name 5-[2-ethoxy-5-(4-methyl-4-oxidopiperazin-4-ium-1-yl)sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CC[N+](C)([O-])CC4

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H30N6O5S/c1-5-7-17-19-20(26(3)25-17)22(29)24-21(23-19)16-14-15(8-9-18(16)33-6-2)34(31,32)27-10-12-28(4,30)13-11-27/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,29)
Formula Weight 490.1998
Percent Purity >95%
Chemical Name or Material Sildenafil N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.