missing translation for 'onlineSavingsMsg'
Learn More

Sildenafil N-Oxide 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30460014
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460014 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460014 Supplier LoGiCal Supplier No. LGCAMP0254.0401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Sildenafil N-Oxide 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sildenafil N-Oxide
CAS 1094598-75-0
Analyte or Component Names Sildenafil N-Oxide
Shipping Condition Room Temperature
Molecular Formula C22 H30 N6 O5 S
Certifications/Compliance ISO 17034
Synonym Sildenafil Citrate Imp. B (EP), Sildenafil Imp. B (EP), 5-[2-Ethoxy-5-[(4-methyl-4-oxidopiperazin-1-yl)sulfonyl]phenyl]-1-methyl-3-propyl-6,7-dihydro-1 Hpyrazolo[4,3-d]pyrimidin-7-one, Sildenafil N-Oxide, Sildenafil Citrate Impurity B, Sildenafil Impurity B
InChI Formula InChI=1 S/C22H30N6O5S/c1-5-7-17-19-20(26(3)25-17)22(29)24-21(23-19)16-14-15(8-9-18(16)33-6-2)34(31,32)27-10-12-28(4,30)13-11-27/h8-9,14 H,5-7,10-13H2,1-4H3,(H,23,24,29)
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CC[N+](C)([O-])CC4
IUPAC Name 5-[2-ethoxy-5-(4-methyl-4-oxidopiperazin-4-ium-1-yl)sulfonylphenyl]-1-methyl-3-propyl-6 H-pyrazolo[4,3-d]pyrimidin-7-one
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 490.58
Quantity 1 mL
Formula Weight 490.1998 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.