missing translation for 'onlineSavingsMsg'
Learn More

Sildenafil, MedChemExpress

Product Code. 16426626
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16426626

Brand: Medchem Express HY15025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sildenafil (UK-92480) is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM.

Sildenafil (UK-92480) is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM.

Specifications

Chemical Name or Material Sildenafil
CAS 139755-83-2
Color White
Purity Grade Notes Research
Molecular Formula C22H30N6O4S
Quantity 100 mg
Synonym UK-92480
Solubility Information DMSO : ≥ 29 mg/mL (61.11 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1C(N(C)N=C2CCC)=C2N=C(C3=CC(S(=O)(N4CCN(C)CC4)=O)=CC=C3OCC)N1
Molecular Weight (g/mol) 474.58
Formula Weight 474.58
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.