missing translation for 'onlineSavingsMsg'
Learn More

Sildenafil, MedChemExpress

Product Code. 16471156
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471156 200 mg 200mg
16426626 100 mg 100mg
16422417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471156 Supplier Medchem Express Supplier No. HY15025
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Sildenafil (UK-92480) is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM.

Sildenafil (UK-92480) is a potent phosphodiesterase type 5 (PDE5) inhibitor with an IC50 of 5.22 nM.

Specifications

Chemical Name or Material Sildenafil
CAS 139755-83-2
Color White
Purity Grade Notes Research
Molecular Formula C22H30N6O4S
Quantity 200 mg
Synonym UK-92480
Solubility Information DMSO : ≥ 29 mg/mL (61.11 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C1C(N(C)N=C2CCC)=C2N=C(C3=CC(S(=O)(N4CCN(C)CC4)=O)=CC=C3OCC)N1
Molecular Weight (g/mol) 474.58
Formula Weight 474.58
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.