missing translation for 'onlineSavingsMsg'
Learn More

BMS-599626 Hydrochloride, MedChemExpress

Product Code. 16481157
Encompass_Preferred
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481157

Brand: Medchem Express HY12010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-599626 Hydrochloride (AC480 Hydrochloride) is a selective and orally bioavailable HER1 and HER2 inhibitor, with IC50s of 20 and 30 nM, respectively. BMS-599626 Hydrochloride displays ∼8-fold less potent to HER4 (IC50=190 nM), >100-fold to VEGFR2, c-Kit, Lck, MEK. BMS-599626 Hydrochloride inhibits tumor cell proliferation, and has potential to increase tumor response to radiotherapy.

BMS-599626 Hydrochloride (AC480 Hydrochloride) is a selective and orally bioavailable HER1 and HER2 inhibitor, with IC50s of 20 and 30 nM, respectively. BMS-599626 Hydrochloride displays ∼8-fold less potent to HER4 (IC50=190 nM), >100-fold to VEGFR2, c-Kit, Lck, MEK. BMS-599626 Hydrochloride inhibits tumor cell proliferation, and has potential to increase tumor response to radiotherapy.

Specifications

Chemical Name or Material BMS-599626 Hydrochloride
CAS 873837-23-1
Color White
Purity Grade Notes Research
Molecular Formula C27H28ClFN8O3
Quantity 50 mg
Synonym AC480 Hydrochloride
Solubility Information DMSO : 100 mg/mL (176.36 mM; Need ultrasonic)
SMILES O=C(OC[C@H]1NCCOC1)NC2=CN(C3=C2C)N=CN=C3NC4=CC5=C(C=C4)N(CC6=CC(F)=CC=C6)N=C5.Cl
Molecular Weight (g/mol) 567.01
Formula Weight 567.01
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.