missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sildenafil

Product Code. 15872534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15872534 1 g 1g
15882534 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15872534 Supplier Thermo Scientific Chemicals Supplier No. 463240010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 139755-83-2 | C22H30N6O4S | 474.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139755-83-2
Molecular Formula C22H30N6O4S
Molecular Weight (g/mol) 474.58
InChI Key BNRNXUUZRGQAQC-UHFFFAOYSA-N
IUPAC Name 5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1

Specifications

Melting Point 188.0°C to 190.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol & dimethylsulfoxide
Formula Weight 474.58
Chemical Name or Material Sildenafil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.