missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ SILBERBRAND Compact Automatic Burette Spare Amber Burette Tube
SILBERBRAND Spare Burette Tube is suitable for use with a compact automatic burette.
£66.50
Specifications
Color | Amber |
---|---|
For Use With (Equipment) | Compact Automatic Burette |
Material | Borosilicate Glass (5.4) |
Type | Buret Tube |
Description
- Constructed of Borosilicate Glass 3.3
- Amber glass
Specifications
Amber | |
Compact Automatic Burette | |
Borosilicate Glass (5.4) | |
Buret Tube |