Learn More
BRAND™ SilberBrand™ Burette with Lateral Stopcock
Product Code. 15211329 Shop All Brand Products
Change view
Click to view available options
Material:
Borosilicate Glass (3.3)
Unit Size:
Pack of 2
Product Code. Material Unit Size
15211329 Borosilicate Glass (3.3) Pack of 2
1 options
Product Code. 15211329 Supplier BRAND™ Supplier No. 13568
  • ONLINE EXCLUSIVE PRICE
    £165.00
    £156.20 / Pack of 2
    Save £8.80
    5% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Boro 3.3 glass class B burette with lateral stopcock
  • Boro 3.3 Glass
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • Black Schellbach stripe
  • Interchangeable needle-valve and glass key
  • Length approx. 800 mm
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description 50 mL volume with 0.1 mL subdivisions
    Material Borosilicate Glass (3.3)
    No. per Pack 2