missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ SILAC RPMI 1640 Medium

Product Code. 15477983
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15477983

Brand: Gibco™ A2494401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SILAC RPMI 1640 Medium

SILAC RPMI 1640 Medium is RPMI 1640 basal cell culture medium without L-arginine, L-glutamine, and L-lysine for use during SILAC protein labeling with stable isotope-labeled lysine and/or arginine. RPMI 1640 Medium is widely used for supporting the growth of many different mammalian cells. Cells successfully cultured in RPMI-1640 include HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of applications. Find the right modification using the media selector tool.

RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). SILAC protein labeling experiments should be conducted using Gibco™ Dialyzed FBS. RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

cGMP Manufacturing and Quality System
SILAC RPMI 1640 Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type RPMI 1640
With Additives Glucose, Phenol Red
Without Additives No Glutamine, No HEPES, No Arginine, No Lysine
For Use With (Application) Mammalian cell culture, primary cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 2 to 8°C
Description SILAC RPMI 1640 Medium, 500mL, Liquid, High Glucose, Phenol Red Indicator, With Glucose, Phenol Red, 12 Months Shelf Life, Standard Serum Supplementation, For Mammalian Cell Culture, Primary Cell Culture

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.