missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24110-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized.Specifications
| SIL1 | |
| Polyclonal | |
| Unconjugated | |
| SIL1 | |
| BAP, BiP associated protein, MSS, SIL1, ULG5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64374, 81500 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9EPK6, Q9H173 | |
| SIL1 | |
| SIL1 Fusion Protein Ag21367 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction