missing translation for 'onlineSavingsMsg'
Learn More

SIL SVOC Mixture 539 200 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30322297
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322297 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322297 Supplier Dr. Ehrenstorfer Supplier No. DREA50000539AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chrysene D12∣Phenanthrene D10∣Pyrene-d10
CAS 1719-03-5, 1517-22-2, 1718-52-1
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Chrysene D12 200 μg/mL∣Phenanthrene D10 200 μg/mL∣Pyrene-d10 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.