missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115974
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total STIL.
Plays an essential role in early embryonic development.
Specifications
| SIL | |
| Polyclonal | |
| Unconjugated | |
| STIL | |
| cassiopeia; chunp6907; csp; MCPH7; nbb; scl/tal1 interrupting locus; SCL-interrupting locus protein; SCL-interrupting locus protein homolog; SIL; sill; Stil; STIL centriolar assembly protein; stil protein; TAL1 (SCL) interrupting locus like; Tal1 interrupting locus; TAL1/SCL interrupting locus-like protein; TAL-1-interrupting locus protein; zgc:110502 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20460, 313506, 6491 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q15468, Q60988 | |
| STIL | |
| A synthesized peptide derived from human STIL(Accession Q15468), corresponding to amino acid residues E1236-L1286. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction