missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ sIL-6ST/gp130/CD130 Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHIL6ST

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15495043

  • £544.00 / 1 set
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

The Human Interleukin 6 ST/GP130/CD130(Hu IL-6ST) ELISA quantitates Hu IL-6ST in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-6ST. Principle of the method The Human IL-6ST solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications

Specifications

P40189
4 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
3572
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Inflammation
2°C to 8°C
Human
4 hr. 45 min.
8.23-6000 pg/mL
4pg/mLto 6000pg/mL
Biotin
Growth Factors & Receptors
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Human
Colorimetric Microplate Reader
CD130,CDW130,GP130,IL-6RB
<10%
HRP
RUO
Plasma, 1 μL; Serum, 1 μL; Supernatant, 100 μL
CD130, CDW130, GP130, IL-6RB
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers