Learn More
Invitrogen™ Human SIK3, GST Tag Recombinant Protein
SIK3 (QSK) is a serine/threonine-protein kinase, belongs to QIK subfamily. Recombinant human SIK3 (1-307) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Brand: Invitrogen™ PV6404
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The phosphorylation of SIK3 by LKB1 through the 14-3-3 binding enhances its catalytic activity and leads its localization to punctate structures within the cytoplasm. Overexpression of SIK3 promotes G1/S cell cycle progression with ovarian cancer. There are two sites (H331L and A1103V) were mutated at significant frequency in breast cancer. SIK3 is a novel tumor-associated antigen (TAA).
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Kinase Assay | |
100μg | |
aa 59-365 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Baculovirus | |
Recombinant, Fragment | |
Serine/Threonine Kinases |
61.7 kDa | |
QSK, SIK-3, L19, FLJ12240, KIAA0999 | |
Human | |
GST Tag | |
Serine/Threonine Kinase | |
Kinases & Inhibitors | |
Liquid |