missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561672
This item is not returnable.
View return policy
Description
Immunogen sequence: QQEYQELFRH MNQGDAGSLA PSLGGQSMTE RQALSYQNAD SYHHHTSPQH LLQIRAQECV SQASSPTPPH GYAHQPALMH SESMEEDCSC EGAKDGFQD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
SIK3 (QSK) is a serine/threonine-protein kinase, belongs to QIK subfamily. The phosphorylation of SIK3 by LKB1 through the 14-3-3 binding enhances its catalytic activity and leads its localization to punctate structures within the cytoplasm. Overexpression of SIK3 promotes G1/S cell cycle progression with ovarian cancer. There are two sites (H331L and A1103V) were mutated at significant frequency in breast cancer. SIK3 is a novel tumor-associated antigen (TAA). SIK3 is a positive regulator of mTOR signaling that functions by triggering the degradation of DEPTOR, an mTOR inhibitor. Involved in the dynamic regulation of mTOR signaling in chondrocyte differentiation during skeletogenesis. Negatively regulates cAMP signaling pathway possibly by acting on CRTC2/TORC2 and CRTC3/TORC3.
Specifications
| SIK3 | |
| Polyclonal | |
| Unconjugated | |
| SIK3 | |
| 5730525O22Rik; 9030204A07Rik; AI447252; AI846254; AI849445; Kiaa0999; L19; mKIAA0999; QSK; Salt-inducible kinase 3; serine/threonine-protein kinase QSK; Serine/threonine-protein kinase SIK3; SIK family kinase 3; SIK3; SIK-3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23387 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2K2 | |
| SIK3 | |
| Recombinant protein corresponding to Human SIK3. Recombinant protein control fragment (Product #RP-100420). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction