missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597005
This item is not returnable.
View return policy
Description
Immunogen sequence: PPPPPPRQPG AAPAPLQFSY QTCELPSAAS PAPDYPTPCQ YPVDGAQQSD LTGPDCPRSP GLQEAPSSYD PLALSELPGL FDCEMLDAVD PQHNGYVLVN; Positive Samples: U-87MG, SW620, A-549, HeLa, Jurkat, HepG2, Mouse fat, Mouse testis, Rat testis; Cellular Location: Cytoplasm.
SNF1LK2 (QIK) is a serine/threonine kinase and is a salt-inducible kinase that belongs to the AMP-activated protein kinase family. SNF1LK2 negatively regulates CRE-binding protein (CREB) activity by phosphorylating the CREB-specific coactivator transducer of regulated CREB activity (TORC). SIK2 is thought to be part of a signaling cascade that regulates the expression and activity of the insulin-induced genes PGC-1'alpha and UCP-1 in brown adipocytes, impairment of which has been implicated in obesity and insulin resistance in human and animal models. SIK2 has also been reported as a key regulator for neuronal survival after ischemia, suppressing CREB-mediated gene expression after oxygen-glucose deprivation.
Specifications
| SIK2 | |
| Polyclonal | |
| Unconjugated | |
| SIK2 | |
| G630080D20Rik; KIAA0781; LOH11CR1I; QIK; Qin-induced kinase; RGD1559698; salt induceable kinase 2; salt inducible kinase 2; salt-inducible kinase 2; salt-inducible protein kinase 2; salt-inducible serine/threonine kinase 2; serine/threonine-protein kinase SIK2; Serine/threonine-protein kinase SNF1-like kinase 2; Sik2; SIK-2; SNF1-like kinase 2; SNF1LK2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23235, 235344, 315649 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.93 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CFH6, Q9H0K1 | |
| SIK2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 827-926 of human SIK2 (NP_0560061). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction