missing translation for 'onlineSavingsMsg'
Learn More

SIK1/Snf1lk Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18409936 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409936 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409936 Supplier Novus Biologicals Supplier No. NBP176571MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SIK1/Snf1lk Polyclonal antibody specifically detects SIK1/Snf1lk in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIK1/Snf1lk
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, msk, myocardial SNF1-like kinase, salt-inducible kinase 1, Salt-inducible protein kinase 1, serine/threonine protein kinase, serine/threonine-protein kinase SIK1, Serine/threonine-protein kinase SNF1-like kinase 1, Serine/threonine-protein kinase SNF1LK, SIK, SIK-1, SNF1-like kinase, SNF1LKMSK
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human SIK1. The immunogen is located within amino acids 340 - 390 of SIK1. Amino Acid Squence: ARPGPARQPRPRSSDLS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 150094
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.