missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SIK1, GST Tag Recombinant Protein

Product Code. 15479053
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 15479053

Brand: Invitrogen™ PV6446

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This recombinant human SIK1 (1-303) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Learn more about our kinase proteins > Related Links:LanthaScreen™ Kinase Activity AssayLanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Active, Catalytic The gene accession number for SIK1 is NP_775490.2.

SIK1 may mediate the physiological effects of LKB1, including its tumor suppressor function. SIK1 is also involved in signaling by various proteins like STRAD, NUAK1, NUAK2, BRSK1, BRSK2, QIK, QSK, SIK, MARK1, MARK2, MARK3, MARK4, and MELK that are related to AMPK. Activation of SIK1 by phosphorylation on thr322 can lead to an increase in the catalytic activity of sodium/potassium ATPase alpha subunit at the plasma membrane. This results in an increase in intracellular sodium in intact mammalian cells. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P57059
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 150mM NaCl, 10mM glutathione, 0.25mM DTT, 25% glycerol, 0.1mM PMSF, 0.1mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 150094
Molecular Weight (g/mol) 61 kDa
Name Human SIK1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias EIEE30; HRT-20; Kid2; MSK; Myocardial SNF1-like kinase; Probable serine/threonine protein kinase SNF1LK (HRT-20) (Myocardial SNF1-like kinase); protein kinase KID2; salt inducible kinase 1; salt-inducible kinase 1; salt-inducible protein kinase 1; serine/threonine protein kinase; serine/threonine-protein kinase SIK1; Serine/threonine-protein kinase SNF1-like kinase 1; serine/threonine-protein kinase SNF1LK; SIK; Sik1; SIK-1; SNF1-like kinase; SNF1LK
Common Name SIK1
Gene Symbol SIK1
Protein Length 1-303
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-303
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Serine/Threonine Kinase
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic Domain
Form Liquid
Research Category Drug Development, Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.